Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTACGTTCCACTGGAGACCTTCAC[A/G]CTGCCCACGCGGTCACTGAAGCCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123631 MIM: 600929 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRYBA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRYBA4 - crystallin beta A4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CRYBB1 - crystallin beta B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001887.3 | 625 | Silent Mutation | AGC,AGT | S,S 185 | NP_001878.1 | |
XM_011529899.2 | 625 | Silent Mutation | AGC,AGT | S,S 185 | XP_011528201.1 |