Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCAGTGGAGGTGTTTGAAGGGG[A/T]GCCAGGGAACAAGGTGAACCTGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601998 MIM: 606583 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESRRA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ESRRA - estrogen related receptor alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRDX5 - peroxiredoxin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012094.4 | 340 | Missense Mutation | GAG,GTG | E,V 71 | NP_036226.1 | |
NM_181651.2 | 340 | Missense Mutation | GAG,GTG | E,V 71 | NP_857634.1 | |
NM_181652.2 | 340 | Intron | NP_857635.1 | |||
XM_005273882.4 | 340 | Intron | XP_005273939.2 |
TRMT112 - tRNA methyltransferase 11-2 homolog (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |