Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACTCCTAAATCAGTTCCAGAGAA[G/T]AATTTATTCAAAGAAGCTTGTGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300348 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLHL4 - kelch like family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019117.4 | 521 | Missense Mutation | AAG,AAT | K,N 113 | NP_061990.2 | |
NM_057162.2 | 521 | Missense Mutation | AAG,AAT | K,N 113 | NP_476503.1 |