Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCTGATCTCAGCATAATGGTAAC[A/G]GATATCCTCTTTCCAACGGGTGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614281 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESAM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ESAM - endothelial cell adhesion molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101929340 - uncharacterized LOC101929340 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MSANTD2 - Myb/SANT DNA binding domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308027.1 | 2378 | Missense Mutation | CGT,TGT | R,C 329 | NP_001294956.1 | |
NM_001312919.1 | 2378 | UTR 3 | NP_001299848.1 | |||
NM_001312920.1 | 2378 | Missense Mutation | CGT,TGT | R,C 99 | NP_001299849.1 | |
NM_001312921.1 | 2378 | Missense Mutation | CGT,TGT | R,C 99 | NP_001299850.1 | |
NM_024631.3 | 2378 | Missense Mutation | CGT,TGT | R,C 277 | NP_078907.2 | |
XM_005271674.3 | 2378 | Intron | XP_005271731.3 | |||
XM_006718905.1 | 2378 | Missense Mutation | CGT,TGT | R,C 99 | XP_006718968.1 | |
XM_006718906.3 | 2378 | Missense Mutation | CGT,TGT | R,C 99 | XP_006718969.1 | |
XM_011542981.2 | 2378 | UTR 3 | XP_011541283.2 | |||
XM_011542982.2 | 2378 | Intron | XP_011541284.2 | |||
XM_017018295.1 | 2378 | Missense Mutation | CGT,TGT | R,C 99 | XP_016873784.1 | |
XM_017018296.1 | 2378 | Missense Mutation | CGT,TGT | R,C 99 | XP_016873785.1 | |
XM_017018297.1 | 2378 | Missense Mutation | CGT,TGT | R,C 99 | XP_016873786.1 | |
XM_017018298.1 | 2378 | Missense Mutation | CGT,TGT | R,C 99 | XP_016873787.1 | |
XM_017018299.1 | 2378 | Intron | XP_016873788.1 | |||
XM_017018300.1 | 2378 | Intron | XP_016873789.1 | |||
XM_017018301.1 | 2378 | Intron | XP_016873790.1 |