Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATAGAAATGTAATGTTAGATAACT[A/G]CAGAAACCTGGTCTTCCTGGGTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF730 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF730 - zinc finger protein 730 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277403.1 | 386 | Intron | NP_001264332.1 | |||
XM_017026114.1 | 386 | Missense Mutation | TAC,TGC | Y,C 48 | XP_016881603.1 | |
XM_017026115.1 | 386 | Missense Mutation | TAC,TGC | Y,C 46 | XP_016881604.1 |