Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCCCCCATCCTTGCCTGTTCCA[C/T]GGCTGTGGCTGGTTTGATGGGGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118503 MIM: 118509 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRNA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHRNA3 - cholinergic receptor nicotinic alpha 3 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CHRNB4 - cholinergic receptor nicotinic beta 4 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000750.3 | 1495 | UTR 3 | NP_000741.1 | |||
NM_001256567.1 | 1495 | Missense Mutation | CAT,CGT | H,R 219 | NP_001243496.1 | |
XM_011521186.2 | 1495 | UTR 3 | XP_011519488.1 | |||
XM_011521187.2 | 1495 | UTR 3 | XP_011519489.1 | |||
XM_011521190.2 | 1495 | UTR 3 | XP_011519492.1 | |||
XM_011521191.2 | 1495 | UTR 3 | XP_011519493.1 | |||
XM_011521192.2 | 1495 | UTR 3 | XP_011519494.1 | |||
XM_017021885.1 | 1495 | UTR 3 | XP_016877374.1 | |||
XM_017021886.1 | 1495 | Intron | XP_016877375.1 | |||
XM_017021887.1 | 1495 | Intron | XP_016877376.1 | |||
XM_017021888.1 | 1495 | Intron | XP_016877377.1 | |||
XM_017021889.1 | 1495 | Intron | XP_016877378.1 |