Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCATATTCATCACAAGACTACTGC[A/G]CTAATGATGGCCTGGCATAAAGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610889 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IPO11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IPO11 - importin 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134779.1 | 1526 | Intron | NP_001128251.1 | |||
NM_016338.4 | 1526 | Intron | NP_057422.3 |
IPO11-LRRC70 - IPO11-LRRC70 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC70 - leucine rich repeat containing 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181506.4 | 1526 | Silent Mutation | GCA,GCG | A,A 429 | NP_852607.3 |