Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCCAGTGGGGGCCAGTGAAATC[A/G]CTGAGGAAACCCGGCCGGGACAACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF853 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF853 - zinc finger protein 853 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017560.1 | 676 | Missense Mutation | ACT,GCT | T,A 79 | NP_060030.1 | |
XM_011515438.2 | 676 | Missense Mutation | ACT,GCT | T,A 105 | XP_011513740.1 | |
XM_011515439.2 | 676 | Missense Mutation | ACT,GCT | T,A 90 | XP_011513741.1 | |
XM_017012371.1 | 676 | Missense Mutation | ACT,GCT | T,A 64 | XP_016867860.1 |