Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGGTCCTGCGCTGCGCCCTGGG[C/G]GACCGCGGCCGCGTGCGGGGCCGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602503 | ||||||||||||||||||||
Literature Links: |
FRAT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FRAT1 - frequently rearranged in advanced T-cell lymphomas 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005479.3 | 537 | Silent Mutation | GGC,GGG | G,G 116 | NP_005470.2 |