Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGCGGACTTCTTCTCCCACTACG[A/T]GTACCTGTGCGCGCTGCAGAACTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP78 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP78 - centrosomal protein 78 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098802.1 | 348 | Missense Mutation | GAG,GTG | E,V 20 | NP_001092272.1 | |
NM_032171.1 | 348 | Missense Mutation | GAG,GTG | E,V 20 | NP_115547.1 | |
XM_005252263.4 | 348 | Missense Mutation | GAG,GTG | E,V 20 | XP_005252320.1 | |
XM_005252265.2 | 348 | Missense Mutation | GAG,GTG | E,V 20 | XP_005252322.1 | |
XM_005252266.4 | 348 | UTR 5 | XP_005252323.1 | |||
XM_017015192.1 | 348 | Missense Mutation | GAG,GTG | E,V 20 | XP_016870681.1 | |
XM_017015193.1 | 348 | Missense Mutation | GAG,GTG | E,V 20 | XP_016870682.1 | |
XM_017015194.1 | 348 | Missense Mutation | GAG,GTG | E,V 20 | XP_016870683.1 | |
XM_017015195.1 | 348 | Missense Mutation | GAG,GTG | E,V 20 | XP_016870684.1 | |
XM_017015196.1 | 348 | UTR 5 | XP_016870685.1 |