Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCGCGAGGAGCGCGCCGGGTGCC[G/T]TCGGGGGGCCTCCGCGCGCGCCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609396 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHLPP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PHLPP1 - PH domain and leucine rich repeat protein phosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_194449.3 | 1116 | Missense Mutation | GTC,TTC | V,F 291 | NP_919431.2 | |
XM_011525886.1 | 1116 | Missense Mutation | GTC,TTC | V,F 291 | XP_011524188.1 |