Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607939 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUMF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SUMF1 - sulfatase modifying factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164674.1 | Intron | NP_001158146.1 | ||||
NM_001164675.1 | Intron | NP_001158147.1 | ||||
NM_182760.3 | Intron | NP_877437.2 | ||||
XM_011533624.2 | Intron | XP_011531926.1 | ||||
XM_011533625.2 | Intron | XP_011531927.1 | ||||
XM_011533626.2 | Intron | XP_011531928.1 | ||||
XM_017006252.1 | Intron | XP_016861741.1 | ||||
XM_017006253.1 | Intron | XP_016861742.1 | ||||
XM_017006254.1 | Intron | XP_016861743.1 | ||||
XM_017006255.1 | Intron | XP_016861744.1 |