Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAATCCACTAGTGACCTAGACCTT[C/T]GATACAGGAGATCCTATAAAACAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606098 | ||||||||||||||||||||
Literature Links: |
SNX6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SNX6 - sorting nexin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021249.3 | 1084 | Missense Mutation | CAA,CGA | Q,R 196 | NP_067072.3 | |
NM_152233.2 | 1084 | Missense Mutation | CAA,CGA | Q,R 324 | NP_689419.2 | |
XM_006720224.1 | 1084 | Missense Mutation | CAA,CGA | Q,R 287 | XP_006720287.1 |