Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTTCTTTGTCTCTGGCGGTAATA[C/T]CTGCTCCGGCTCTGGCTGCGACAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182880 MIM: 182890 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRM1 - protamine 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002761.2 | 139 | Silent Mutation | AGA,AGG | R,R 14 | NP_002752.1 |
PRM2 - protamine 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRM3 - protamine 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |