Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCACCAAGCTGAATGCCAGGATGG[A/G]GTCCACTCTCTGCACCTCACCTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614313 MIM: 611919 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACOT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACOT1 - acyl-CoA thioesterase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037161.1 | 1320 | Intron | NP_001032238.1 | |||
XM_017021589.1 | 1320 | Intron | XP_016877078.1 | |||
XM_017021590.1 | 1320 | Intron | XP_016877079.1 | |||
XM_017021591.1 | 1320 | Intron | XP_016877080.1 | |||
XM_017021592.1 | 1320 | Intron | XP_016877081.1 |
C14orf169 - chromosome 14 open reading frame 169 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024644.4 | 1320 | Silent Mutation | GGA,GGG | G,G 412 | NP_078920.2 |
HEATR4 - HEAT repeat containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001220484.1 | 1320 | Intron | NP_001207413.1 | |||
NM_203309.2 | 1320 | Intron | NP_976054.2 | |||
XM_006720143.2 | 1320 | Intron | XP_006720206.1 | |||
XM_011536760.2 | 1320 | Intron | XP_011535062.1 | |||
XM_011536761.2 | 1320 | Intron | XP_011535063.1 | |||
XM_017021289.1 | 1320 | Intron | XP_016876778.1 |