Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCTTTCCCCTCTCTCACGTTGC[A/G]GGAGTCCTTCTGGTCTTGCCCTCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603767 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLK4 - kallikrein related peptidase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302961.1 | 607 | Missense Mutation | CGC,TGC | R,C 108 | NP_001289890.1 | |
NM_004917.4 | 607 | Missense Mutation | CGC,TGC | R,C 203 | NP_004908.4 | |
XM_005259441.4 | 607 | Missense Mutation | CGC,TGC | R,C 108 | XP_005259498.2 | |
XM_011527545.2 | 607 | Intron | XP_011525847.1 | |||
XM_011527546.2 | 607 | Intron | XP_011525848.1 |