Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCAGCTCTTGCTCGGTTGGGAGA[C/T]GGTGCAAGAGAATCTGCCCCCTATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605994 MIM: 613725 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP39A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP39A1 - cytochrome P450 family 39 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A27 - solute carrier family 25 member 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204051.1 | 458 | Silent Mutation | GAC,GAT | D,D 60 | NP_001190980.1 | |
NM_001204052.1 | 458 | Silent Mutation | GAC,GAT | D,D 60 | NP_001190981.1 | |
NM_004277.4 | 458 | Silent Mutation | GAC,GAT | D,D 60 | NP_004268.3 | |
XM_005249483.2 | 458 | Silent Mutation | GAC,GAT | D,D 60 | XP_005249540.1 | |
XM_017011512.1 | 458 | UTR 5 | XP_016867001.1 |