Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATAGGTAATTTTTTGTGTTCTTC[A/T]TGCTATCAGAAGCTGACCAATGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614631 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ISPD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ISPD - isoprenoid synthase domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011515499.2 | Intron | XP_011513801.1 | ||||
XM_011515501.2 | Intron | XP_011513803.1 | ||||
XM_017012575.1 | Intron | XP_016868064.1 | ||||
XM_017012576.1 | Intron | XP_016868065.1 |