Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTACAAATTCCACTGTTACCCATG[C/T]ATCGAAGGATGTGGCGTTTGCCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607650 MIM: 615997 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEFB118 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DEFB118 - defensin beta 118 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DEFB119 - defensin beta 119 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271209.1 | 249 | Intron | NP_001258138.1 | |||
NM_153289.3 | 249 | Missense Mutation | TAC,TGC | Y,C 28 | NP_695021.2 | |
NM_153323.4 | 249 | Intron | NP_697018.1 |