Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTGGGGCGGCGGAGCCTTCCTCT[C/T]CCTTGACGTCTTGCGAGCTCTTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600724 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNGB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNGB1 - cyclic nucleotide gated channel beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135639.1 | 3553 | Intron | NP_001129111.1 | |||
NM_001286130.1 | 3553 | Missense Mutation | GAA,GGA | E,G 1157 | NP_001273059.1 | |
NM_001297.4 | 3553 | Missense Mutation | GAA,GGA | E,G 1163 | NP_001288.3 | |
XM_011522870.2 | 3553 | Missense Mutation | GAA,GGA | E,G 780 | XP_011521172.1 |