Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCGTGCGCACCCCGCCCGGGCCA[C/T]CCCTGCGCCTGGCGCGCAACGGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600222 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TIE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TIE1 - tyrosine kinase with immunoglobulin like and EGF like domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253357.1 | 323 | Missense Mutation | CCC,TCC | P,S 32 | NP_001240286.1 | |
NM_005424.4 | 323 | Missense Mutation | CCC,TCC | P,S 77 | NP_005415.1 | |
XM_005271163.1 | 323 | Missense Mutation | CCC,TCC | P,S 77 | XP_005271220.1 | |
XM_006710869.1 | 323 | UTR 5 | XP_006710932.1 | |||
XM_017002207.1 | 323 | Missense Mutation | CCC,TCC | P,S 77 | XP_016857696.1 |