Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCTCAGGTTGGAATGCCAGGAA[A/G]TGGACCAGTGTCCATGGAACGGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300907 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSTF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSTF2 - cleavage stimulation factor subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306206.1 | 914 | Missense Mutation | AAT,AGT | N,S 287 | NP_001293135.1 | |
NM_001306209.1 | 914 | Missense Mutation | AAT,AGT | N,S 270 | NP_001293138.1 | |
NM_001325.2 | 914 | Missense Mutation | AAT,AGT | N,S 287 | NP_001316.1 |