Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTCCGGCCCCCGCCATGGCCCG[C/G]GGCGGCAGTCAGAGCTGGAGCTCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
DNAJA4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAJA4 - DnaJ heat shock protein family (Hsp40) member A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130182.1 | 178 | Intron | NP_001123654.1 | |||
NM_001130183.1 | 178 | Intron | NP_001123655.1 | |||
NM_018602.3 | 178 | Silent Mutation | CGC,CGG | R,R 3 | NP_061072.3 | |
XM_005254535.3 | 178 | Intron | XP_005254592.1 | |||
XM_006720601.1 | 178 | Intron | XP_006720664.1 |