Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGTGGCTTTAAACCACCAAAAG[A/G]TGCCTGTTGCTGGCTTTTGAGCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613491 MIM: 613305 MIM: 601119 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACER1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACER1 - alkaline ceramidase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ALKBH7 - alkB homolog 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CLPP - caseinolytic mitochondrial matrix peptidase proteolytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006012.2 | Intron | NP_006003.1 |