Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCGGGGGGCGCCCCAGACGCGCA[A/G]CAGCTGCGCCAGGACGCGCGCCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300437 MIM: 300272 MIM: 300399 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERAS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERAS - ES cell expressed Ras | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HDAC6 - histone deacetylase 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCSK1N - proprotein convertase subtilisin/kexin type 1 inhibitor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013271.4 | 434 | Silent Mutation | CTG,TTG | L,L 110 | NP_037403.1 |