Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTTCTTCGCTGTCTGAGTCCTC[A/G]TATAGGTTGATGGTTGCCTGGACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300520 MIM: 300575 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLDN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLDN2 - claudin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171092.1 | 437 | Intron | NP_001164563.1 | |||
NM_001171095.1 | 437 | Intron | NP_001164566.1 | |||
NM_020384.3 | 437 | Intron | NP_065117.1 |
RIPPLY1 - ripply transcriptional repressor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171706.1 | 437 | Silent Mutation | TAC,TAT | Y,Y 84 | NP_001165177.1 | |
NM_138382.2 | 437 | Silent Mutation | TAC,TAT | Y,Y 131 | NP_612391.1 |