Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCGCTGACCTGGGGAAGCAGTGC[A/G]CCGCTGTCAGCACCCACTGGGGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613797 MIM: 600787 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRSS33 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRSS33 - protease, serine 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152891.2 | 323 | Missense Mutation | NP_690851.2 | |||
XM_011522451.2 | 323 | Missense Mutation | XP_011520753.1 | |||
XM_011522452.2 | 323 | Missense Mutation | XP_011520754.1 |
PRSS41 - protease, serine 41 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TCEB2 - transcription elongation factor B subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |