Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAGATGATGCTTGCTTCTGCCCC[A/C]ACAGTGTATCCCTTCTTCAGACTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123260 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRP - C-reactive protein, pentraxin-related | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000567.2 | 539 | Silent Mutation | NP_000558.2 | |||
XM_011509207.2 | 539 | Silent Mutation | XP_011507509.1 |