Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCGCTTTGCGTCCTGGGACGAGAT[C/G]AATGTCCTGGCGCACGGACTCCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605910 | ||||||||||||||||||||
Literature Links: |
ANGPTL4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANGPTL4 - angiopoietin like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039667.2 | 318 | Missense Mutation | ATC,ATG | I,M 41 | NP_001034756.1 | |
NM_139314.2 | 318 | Missense Mutation | ATC,ATG | I,M 41 | NP_647475.1 | |
XM_005272484.3 | 318 | Missense Mutation | ATC,ATG | I,M 41 | XP_005272541.1 | |
XM_005272485.3 | 318 | Missense Mutation | ATC,ATG | I,M 41 | XP_005272542.1 |
RAB11B-AS1 - RAB11B antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |