Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCAGTCCATCTGAGGCACAGCAA[A/G]TTGCATAACATCATTACCACTTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114106 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP3CB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP3CB - protein phosphatase 3 catalytic subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142353.2 | 1638 | Intron | NP_001135825.1 | |||
NM_001142354.2 | 1638 | Intron | NP_001135826.1 | |||
NM_001289968.1 | 1638 | Missense Mutation | CTT,TTT | L,F 480 | NP_001276897.1 | |
NM_001289969.1 | 1638 | Missense Mutation | CTT,TTT | L,F 376 | NP_001276898.1 | |
NM_021132.3 | 1638 | Intron | NP_066955.1 | |||
XM_005269944.1 | 1638 | Intron | XP_005270001.1 | |||
XM_017016379.1 | 1638 | Intron | XP_016871868.1 |