Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAATGCATTTCTCATCATTTACA[C/G]TATGATTGTCAGGATTTGGCTGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611923 MIM: 606535 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GJA9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GJA9 - gap junction protein alpha 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030772.4 | 1231 | Missense Mutation | ACT,AGT | T,S 314 | NP_110399.2 |
GJA9-MYCBP - GJA9-MYCBP readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105378663 - uncharacterized LOC105378663 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYCBP - MYC binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |