Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGAGGAGGCACCCCCTACCCGGG[C/T]CATTTCTACCAAGGCCCTGAGGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610837 MIM: 603734 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL2L12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCL2L12 - BCL2 like 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IRF3 - interferon regulatory factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197122.1 | 1446 | Missense Mutation | GAC,GGC | D,G 384 | NP_001184051.1 | |
NM_001197123.1 | 1446 | Missense Mutation | ACC,GCC | T,A 344 | NP_001184052.1 | |
NM_001197124.1 | 1446 | Missense Mutation | ACC,GCC | T,A 252 | NP_001184053.1 | |
NM_001197125.1 | 1446 | Missense Mutation | ACC,GCC | T,A 233 | NP_001184054.1 | |
NM_001197126.1 | 1446 | Missense Mutation | ACC,GCC | T,A 233 | NP_001184055.1 | |
NM_001197127.1 | 1446 | Missense Mutation | ACC,GCC | T,A 106 | NP_001184056.1 | |
NM_001197128.1 | 1446 | Missense Mutation | ACC,GCC | T,A 106 | NP_001184057.1 | |
NM_001571.5 | 1446 | Missense Mutation | ACC,GCC | T,A 379 | NP_001562.1 | |
XM_006723197.1 | 1446 | Missense Mutation | GAC,GGC | D,G 384 | XP_006723260.1 | |
XM_006723198.1 | 1446 | Missense Mutation | GAC,GGC | D,G 384 | XP_006723261.1 | |
XM_006723200.1 | 1446 | Missense Mutation | GAC,GGC | D,G 349 | XP_006723263.1 | |
XM_006723201.1 | 1446 | Missense Mutation | GAC,GGC | D,G 238 | XP_006723264.1 | |
XM_006723202.2 | 1446 | Missense Mutation | GAC,GGC | D,G 238 | XP_006723265.1 | |
XM_017026766.1 | 1446 | Missense Mutation | ACC,GCC | T,A 379 | XP_016882255.1 | |
XM_017026767.1 | 1446 | Missense Mutation | ACC,GCC | T,A 379 | XP_016882256.1 |
SCAF1 - SR-related CTD associated factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |