Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTGTGGATGTGGTGGCGCAGGA[A/C]CCGCCGCGGGAAGCTTGGGCGCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616692 | ||||||||||||||||||||
Literature Links: |
ESYT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ESYT3 - extended synaptotagmin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322831.1 | 450 | Missense Mutation | AAC,ACC | N,T 75 | NP_001309760.1 | |
NM_001322834.1 | 450 | Missense Mutation | AAC,ACC | N,T 75 | NP_001309763.1 | |
NM_031913.4 | 450 | Missense Mutation | AAC,ACC | N,T 75 | NP_114119.2 | |
XM_017007307.1 | 450 | Intron | XP_016862796.1 |