Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATGCACAGTTTGTACCCGGGTTT[A/G]CCAGTGTAGAATCCAGGGCTATGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602355 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAF6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRAF6 - TNF receptor associated factor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004620.3 | 1437 | Silent Mutation | GGC,GGT | G,G 383 | NP_004611.1 | |
NM_145803.2 | 1437 | Silent Mutation | GGC,GGT | G,G 383 | NP_665802.1 | |
XM_017018220.1 | 1437 | Intron | XP_016873709.1 |