Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGTCATAGTGGCCGATGATGGCA[C/T]TGGCCACTTGCAAGGCCACATTGTC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 120120 MIM: 191328 | |||||||||||||||||||||||
Literature Links: |
COL7A1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
COL7A1 - collagen type VII alpha 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA94 - small nucleolar RNA, H/ACA box 94 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UQCRC1 - ubiquinol-cytochrome c reductase core protein I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003365.2 | 968 | Missense Mutation | NP_003356.2 |