Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTATAATCTAAACATTCAGACGA[A/G]ATCCCTCTACTGCGAATAGCCCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603565 MIM: 614784 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNT4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GALNT4 - polypeptide N-acetylgalactosaminyltransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003774.4 | 1616 | Silent Mutation | ATC,ATT | I,I 453 | NP_003765.2 |
POC1B - POC1 centriolar protein B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199777.1 | 1616 | Intron | NP_001186706.1 | |||
NM_172240.2 | 1616 | Intron | NP_758440.1 |
POC1B-GALNT4 - POC1B-GALNT4 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199781.1 | 1616 | Silent Mutation | ATC,ATT | I,I 450 | NP_001186710.1 | |
NM_001199782.1 | 1616 | Silent Mutation | ATC,ATT | I,I 281 | NP_001186711.1 |