Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCACCTAGGGGGCGGCTACCCC[A/G]TGGGGTAAGTGACTGCAGCTATGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR44 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WDR44 - WD repeat domain 44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184965.1 | 504 | Missense Mutation | ATG,GTG | M,V 25 | NP_001171894.1 | |
NM_001184966.1 | 504 | Missense Mutation | ATG,GTG | M,V 25 | NP_001171895.1 | |
NM_019045.4 | 504 | Missense Mutation | ATG,GTG | M,V 25 | NP_061918.3 | |
XM_011531353.2 | 504 | Missense Mutation | ATG,GTG | M,V 25 | XP_011529655.1 | |
XM_017029600.1 | 504 | UTR 5 | XP_016885089.1 |