Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTGTGGAAAATCTTGACAACAT[C/T]AGAAAGGTGCGAAAAAAGGTGAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADNP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADNP2 - ADNP homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014913.3 | 271 | Silent Mutation | ATC,ATT | I,I 12 | NP_055728.1 | |
XM_005266656.4 | 271 | Silent Mutation | ATC,ATT | I,I 12 | XP_005266713.1 | |
XM_011525883.2 | 271 | Silent Mutation | ATC,ATT | I,I 12 | XP_011524185.1 |