Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACTTATGGGCATAAAAACCTGGG[A/C]CTCTGGGTTAAAGCTGTTTTTGATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605523 | ||||||||||||||||||||
Literature Links: |
TOB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TOB1 - transducer of ERBB2, 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243877.1 | 646 | Missense Mutation | GCC,TCC | A,S 136 | NP_001230806.1 | |
NM_001243885.1 | 646 | UTR 5 | NP_001230814.1 | |||
NM_005749.3 | 646 | Missense Mutation | GCC,TCC | A,S 136 | NP_005740.1 |
TOB1-AS1 - TOB1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |