Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTAGACACTGGTGGCCACCACG[C/T]GGAATTATTAAGGTAATGTTCCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPINK14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPINK14 - serine peptidase inhibitor, Kazal type 14 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001325.2 | 266 | Silent Mutation | CGC,CGT | R,R 33 | NP_001001325.1 | |
XM_017009466.1 | 266 | Silent Mutation | CGC,CGT | R,R 50 | XP_016864955.1 | |
XM_017009467.1 | 266 | Intron | XP_016864956.1 | |||
XM_017009468.1 | 266 | Silent Mutation | CGC,CGT | R,R 26 | XP_016864957.1 | |
XM_017009469.1 | 266 | Silent Mutation | CGC,CGT | R,R 33 | XP_016864958.1 |