Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGGCCCCCATGGGGGGCCGGCCG[A/G]GAGCGGCAGCCGGTGCCTCCTCGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM155B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM155B - family with sequence similarity 155 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015686.2 | 306 | Silent Mutation | CGA,CGG | R,R 86 | NP_056501.2 | |
XM_011530908.2 | 306 | Silent Mutation | CGA,CGG | R,R 86 | XP_011529210.1 | |
XM_011530909.2 | 306 | Silent Mutation | CGA,CGG | R,R 86 | XP_011529211.1 |