Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGCTCTGACTCGGGAAACAGCTC[C/T]TCTTCTTCCTTCTCCTTCTTGGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604819 MIM: 607733 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR937 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR937 - microRNA 937 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PUF60 - poly(U) binding splicing factor 60 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136033.2 | 1349 | Silent Mutation | GAA,GAG | E,E 379 | NP_001129505.1 | |
NM_001271096.1 | 1349 | Silent Mutation | GAA,GAG | E,E 404 | NP_001258025.1 | |
NM_001271097.1 | 1349 | Silent Mutation | GAA,GAG | E,E 376 | NP_001258026.1 | |
NM_001271098.1 | 1349 | Silent Mutation | GAA,GAG | E,E 421 | NP_001258027.1 | |
NM_001271099.1 | 1349 | Silent Mutation | GAA,GAG | E,E 393 | NP_001258028.1 | |
NM_001271100.1 | 1349 | Silent Mutation | GAA,GAG | E,E 362 | NP_001258029.1 | |
NM_014281.4 | 1349 | Silent Mutation | GAA,GAG | E,E 405 | NP_055096.2 | |
NM_078480.2 | 1349 | Silent Mutation | GAA,GAG | E,E 422 | NP_510965.1 | |
XM_011516929.2 | 1349 | Silent Mutation | GAA,GAG | E,E 459 | XP_011515231.1 | |
XM_011516930.2 | 1349 | Silent Mutation | GAA,GAG | E,E 442 | XP_011515232.1 | |
XM_017013234.1 | 1349 | Silent Mutation | GAA,GAG | E,E 459 | XP_016868723.1 | |
XM_017013235.1 | 1349 | Silent Mutation | GAA,GAG | E,E 447 | XP_016868724.1 | |
XM_017013236.1 | 1349 | Silent Mutation | GAA,GAG | E,E 442 | XP_016868725.1 | |
XM_017013237.1 | 1349 | Silent Mutation | GAA,GAG | E,E 459 | XP_016868726.1 | |
XM_017013238.1 | 1349 | Silent Mutation | GAA,GAG | E,E 459 | XP_016868727.1 | |
XM_017013239.1 | 1349 | Silent Mutation | GAA,GAG | E,E 379 | XP_016868728.1 | |
XM_017013240.1 | 1349 | Silent Mutation | GAA,GAG | E,E 362 | XP_016868729.1 |
SCRIB - scribbled planar cell polarity protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |