Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCACCTGGGACCATGTGTCCGAGG[C/T]TGGGGCTTGGGTCGGCCCACACCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601480 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYO1F PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYO1F - myosin IF | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012335.3 | 3180 | Silent Mutation | NP_036467.2 | |||
XM_011528024.1 | 3180 | Missense Mutation | XP_011526326.1 | |||
XM_011528025.1 | 3180 | Missense Mutation | XP_011526327.1 | |||
XM_011528026.1 | 3180 | Missense Mutation | XP_011526328.1 | |||
XM_011528027.1 | 3180 | Missense Mutation | XP_011526329.1 | |||
XM_011528028.2 | 3180 | Intron | XP_011526330.1 | |||
XM_017026821.1 | 3180 | Silent Mutation | XP_016882310.1 |
ZNF414 - zinc finger protein 414 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |