Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATGTGGCCCGATTCACTGATGCT[C/G]CACGTCTGGCGCGGCAGGCGGCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114280 MIM: 609151 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AIM1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AIM1L - absent in melanoma 1-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039775.3 | 4969 | Missense Mutation | TGC,TGG | C,W 1611 | NP_001034864.2 | |
XM_005245918.2 | 4969 | Missense Mutation | TGC,TGG | C,W 1611 | XP_005245975.1 | |
XM_011541672.1 | 4969 | Missense Mutation | TGC,TGG | C,W 1599 | XP_011539974.1 | |
XM_011541673.2 | 4969 | Missense Mutation | TGC,TGG | C,W 1668 | XP_011539975.1 |
CD52 - CD52 molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBXN11 - UBX domain protein 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |