Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGAGAAGAGCCCAGAGTATAGAG[C/T]GGAAGCAAGAGAGGTAAGGAGTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605170 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EI24 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EI24 - EI24, autophagy associated transmembrane protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290135.1 | 448 | Intron | NP_001277064.1 | |||
NM_004879.4 | 448 | Intron | NP_004870.3 | |||
XM_011543069.1 | 448 | Missense Mutation | CGG,TGG | R,W 59 | XP_011541371.1 | |
XM_011543070.1 | 448 | Missense Mutation | CGG,TGG | R,W 59 | XP_011541372.1 | |
XM_017018582.1 | 448 | Missense Mutation | CGG,TGG | R,W 59 | XP_016874071.1 |
STT3A-AS1 - STT3A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |