Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTTCGGACTTGTCTTCGCGGCC[C/T]CAGTCCCCGACCTCGGCGCTGCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHF14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PHF14 - PHD finger protein 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014660.3 | 409 | UTR 5 | NP_055475.2 | |||
XM_017012811.1 | 409 | UTR 5 | XP_016868300.1 | |||
XM_017012812.1 | 409 | UTR 5 | XP_016868301.1 | |||
XM_017012813.1 | 409 | UTR 5 | XP_016868302.1 | |||
XM_017012814.1 | 409 | UTR 5 | XP_016868303.1 | |||
XM_017012815.1 | 409 | UTR 5 | XP_016868304.1 | |||
XM_017012816.1 | 409 | UTR 5 | XP_016868305.1 | |||
XM_017012817.1 | 409 | UTR 5 | XP_016868306.1 |