Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCACAAGCCCCCCGCGGGAAGT[A/G]GGAAATCGACCGACCAGGCGGTGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612766 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKLE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKLE1 - ankyrin repeat and LEM domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278443.1 | 232 | Missense Mutation | AGG,GGG | R,G 40 | NP_001265372.1 | |
NM_001278444.1 | 232 | Missense Mutation | AGG,GGG | R,G 40 | NP_001265373.1 | |
NM_001278445.1 | 232 | Silent Mutation | GTA,GTG | V,V 9 | NP_001265374.1 | |
NM_152363.5 | 232 | Missense Mutation | AGG,GGG | R,G 40 | NP_689576.5 |
BABAM1 - BRISC and BRCA1 A complex member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |