Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCTCATAAATGAGGCCCAAGAT[A/G]CGCTTGACACCGCCATGCCGGGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602806 MIM: 602816 MIM: 602824 MIM: 602832 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIST1H2BH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIST1H2BH - histone cluster 1, H2bh | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H3F - histone cluster 1, H3f | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H4F - histone cluster 1, H4f | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H4G - histone cluster 1, H4g | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003547.2 | 138 | Silent Mutation | CGC,CGT | R,R 46 | NP_003538.1 |