Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCAGAGAGCCCTCTAGTGACCAC[G/A]AAGGGGAGTTAATGCAGAGATGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604323 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCC3 - ATP binding cassette subfamily C member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ANKRD40 - ankyrin repeat domain 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052855.3 | 1821 | UTR 3 | NP_443087.1 | |||
XM_017025296.1 | 1821 | UTR 3 | XP_016880785.1 |