Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACGTACGATACTTAAAACCATCA[C/T]AAGCTGCCCAAGCAATAGAAAACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604961 MIM: 608888 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE11A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDE11A - phosphodiesterase 11A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBM45 - RNA binding motif protein 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152945.3 | 1084 | Nonsense Mutation | CAA,TAA | Q,* 173 | NP_694453.2 | |
XM_005246287.4 | 1084 | Nonsense Mutation | CAA,TAA | Q,* 173 | XP_005246344.1 | |
XM_017003320.1 | 1084 | Nonsense Mutation | CAA,TAA | Q,* 316 | XP_016858809.1 | |
XM_017003321.1 | 1084 | Nonsense Mutation | CAA,TAA | Q,* 316 | XP_016858810.1 | |
XM_017003322.1 | 1084 | Nonsense Mutation | CAA,TAA | Q,* 173 | XP_016858811.1 |